missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2S-cis)-(+)-2,3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one, 98%, ACROS Organics™

$122.68 - $122.68

Chemical Identifiers

CAS 42399-49-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C16H15NO3S
Formula Weight 301.36
MDL Number MFCD01321316
Chemical Name or Material (2S-cis)-(+)-2, 3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1, 5-benzothiazepin-4(5H)-one, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC334220050 SDS Thermo Fisher Scientific
334220050
5g Each for $122.68
Specifications Description & Specifications

Chemical Identifiers

42399-49-5
100.0
C16H15NO3S
MFCD01321316
≥97.5%
97.5
98%
301.36
(2S-cis)-(+)-2, 3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1, 5-benzothiazepin-4(5H)-one, 98%

Specifications

5g
97.5
Authentic
Powder
+110° (114°C )
White to slightly beige
MFCD01321316
(2S-cis)-(+)-2, 3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1, 5-benzothiazepin-4(5H)-one, 98%
42399-49-5
100.0
+110° to +114° (20°C, 589nm) (c=0.5, CH3OH)
98%
C16H15NO3S
301.36
203° to 207°C
≥97.5%

Safety and Handling

EINECSNumber : 255-799-0

TSCA : TSCA