Please login to your online account to display your discounted pricing

(2S,5S)-(+)-Hexanediol, 99%, 99% e.e., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC318760010

 View more versions of this product

Catalog No. AC318760010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystals
Formula Weight 118.18
Quantity 1g
Color White
TSCA TSCA
Specific Rotation Condition + 36.00 (20.00°C c=5,CHCl3)
Melting Point 50° to 53°C
CAS Max % 100.0
Enantiomeric Excess 98.5% min. (GC)
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 34338-96-0
Specific Rotation +35° to +37° (20°C, 589nm) (c=5, CHCl3)
CAS Min % 98.5
Health Hazard 1 HYGROSCOPIC
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material (2S, 5S)-(+)-Hexanediol
Name Note 99% (99% E.E.)
Assay Percent Range 99%
Molecular Formula C6H14O2
Linear Formula CH3CH(OH)CH2CH2CH(OH)CH3
MDL Number MFCD00082583

Specifications

Chemical Name or Material (2S, 5S)-(+)-Hexanediol
Name Note 99% (99% E.E.)
CAS 34338-96-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H14O2
Linear Formula CH3CH(OH)CH2CH2CH(OH)CH3
Formula Weight 118.18
MDL Number MFCD00082583
Accessories

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents