Please login to your online account to display your discounted pricing

(2S,5S)-(+)-Hexanediol, 99%, 99% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (2S, 5S)-(+)-Hexanediol
Name Note 99% (99% E.E.)
CAS 34338-96-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H14O2
Linear Formula CH3CH(OH)CH2CH2CH(OH)CH3
Formula Weight 118.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC318760010

  SDS
thermo fisher scientific
AC318760010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystals
Formula Weight 118.18
Quantity 1g
Color White
TSCA TSCA
Specific Rotation Condition + 36.00 (20.00°C c=5,CHCl3)
Melting Point 50° to 53°C
CAS Max % 100.0
Enantiomeric Excess 98.5% min. (GC)
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 34338-96-0
Specific Rotation +35° to +37° (20°C, 589nm) (c=5, CHCl3)
CAS Min % 98.5
Health Hazard 1 HYGROSCOPIC
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material (2S, 5S)-(+)-Hexanediol
Name Note 99% (99% E.E.)
Assay Percent Range 99%
Molecular Formula C6H14O2
Linear Formula CH3CH(OH)CH2CH2CH(OH)CH3
MDL Number MFCD00082583
Safety and Handling

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.