Please login to your online account to display your discounted pricing

Alfa Aesar™ (2S,5S)-2,5-Hexanediol, 99%

$78.50 - $219.99

Chemical Identifiers

Chemical Name or Material (2S,5S)-2,5-Hexanediol
CAS 34338-96-0
Molecular Formula C6H14O2
Formula Weight 118.18
MDL Number MFCD00082583
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2553901

  SDS
alfa aesar™
B2553901
0.25g Each for $78.50

AAB2553903

  SDS
alfa aesar™
B2553903
1g Each for $219.99
Description & Specifications

Specifications

Formula Weight 118.18
Assay 99%
Boiling Point 216° to 218°C
CAS 34338-96-0
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Quantity 0.25g
Molecular Formula C6H14O2
Melting Point 52° to 54°C
MDL Number MFCD00082583
Flash Point 101°C (215°F)
Chemical Name or Material (2S,5S)-2,5-Hexanediol
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.