missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2S,5S)-1-(2-[(2S,5S)-2,5-Diethyl-1-phospholanyl]phenyl)-2,5-diethylphospholane 1-oxide, 97+%, Alfa Aesar™

Chemical Identifiers

Molecular Formula C22H36OP2
MDL Number MFCD17018781
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60788MD View Documents Alfa Aesar
H60788MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6078803 View Documents Alfa Aesar
H6078803
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

C22H36OP2
MFCD17018781

Specifications

250mg
MFCD17018781
≥97%
C22H36OP2
378.47
(2S,5S)-1-(2-[(2S,5S)-2,5-Diethyl-1-phospholanyl]phenyl)-2,5-diethylphospholane 1-oxide