missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC435175000
Specifications
(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane | |
>110°C | |
96.0% min. | |
1.103 | |
500mg | |
96.0 | |
97% | |
1-2-1,3-dioxolane-2-yl phenyl-2beta,5alpha-dimethyltetrahydro-1h-phosphole, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min, 2-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, kanata purity, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, s,s-me-rajphos | |
CC1CCC(P1C2=CC=CC=C2C3OCCO3)C | |
264.3 | |
264.3 | |
97% |
1.1030g/mL | |
Authentic | |
Glass bottle | |
Colorless to Yellow | |
695816-47-8 | |
100.0 | |
C15H21O2P | |
JAKXTLMNIHPLEU-RYUDHWBXSA-N | |
2-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane | |
16218307 | |
Liquid |
Chemical Identifiers
695816-47-8 | |
264.3 | |
1-2-1,3-dioxolane-2-yl phenyl-2beta,5alpha-dimethyltetrahydro-1h-phosphole, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min, 2-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, kanata purity, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, s,s-me-rajphos | |
2-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane |
C15H21O2P | |
JAKXTLMNIHPLEU-RYUDHWBXSA-N | |
16218307 | |
CC1CCC(P1C2=CC=CC=C2C3OCCO3)C |