missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane, 97%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC435171000

 View more versions of this product

Catalog No. AC435171000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane
96.0
1.1030g/mL
>110°C
Authentic
96.0% min.
1-2-1,3-dioxolane-2-yl phenyl-2beta,5alpha-dimethyltetrahydro-1h-phosphole, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min, 2-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, kanata purity, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, s,s-me-rajphos
JAKXTLMNIHPLEU-RYUDHWBXSA-N
CC1CCC(P1C2=CC=CC=C2C3OCCO3)C
264.3
264.3
97%
695816-47-8
100.0
Colorless to Yellow
97%
C15H21O2P
Glass bottle
100mg
1.103
2-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane
16218307
Liquid

Chemical Identifiers

695816-47-8
264.3
1-2-1,3-dioxolane-2-yl phenyl-2beta,5alpha-dimethyltetrahydro-1h-phosphole, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min, 2-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, kanata purity, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, s,s-me-rajphos
2-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane
C15H21O2P
JAKXTLMNIHPLEU-RYUDHWBXSA-N
16218307
CC1CCC(P1C2=CC=CC=C2C3OCCO3)C
SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only