Please login to your online account to display your discounted pricing

Alfa Aesar™ (2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane, 97%

Chemical Identifiers

Chemical Name or Material (2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane
CAS 695816-47-8
Formula Weight 264.3
MDL Number MFCD09265097
Synonym S,S-Me-RajPhos
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH63462MC

  SDS
alfa aesar™
H63462MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63462ME

  SDS
alfa aesar™
H63462ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 264.3
Chemical Name or Material (2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane
Density 1.103g/mL
Physical Form Liquid
Assay 97%
MDL Number MFCD09265097
CAS 695816-47-8
Synonym S,S-Me-RajPhos
Refractive Index 1.5715
Color Pale yellow to colorless
Sensitivity Air Sensitive
Quantity 100mg