missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC434125000

 View more versions of this product

Catalog No. AC434125000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane
Authentic
Glass bottle
500mg
97%
2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, s,s-et-rajphos, 2-2-2s,5s-2,5-diethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-1,3-dioxolane, trans-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, kanata purity
CCC1CCC(P1C2=CC=CC=C2C3OCCO3)CC
292.359
292.35
97%
>110°C
96.0% min.
Colorless to Yellow
1314246-02-0
C17H25O2P
UKBFKBMQNUOKTD-KBPBESRZSA-N
2-[2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl]-1,3-dioxolane
16218309
Liquid

Chemical Identifiers

1314246-02-0
292.359
2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, s,s-et-rajphos, 2-2-2s,5s-2,5-diethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-1,3-dioxolane, trans-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, kanata purity
2-[2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl]-1,3-dioxolane
C17H25O2P
UKBFKBMQNUOKTD-KBPBESRZSA-N
16218309
CCC1CCC(P1C2=CC=CC=C2C3OCCO3)CC
SDS Documents