Please login to your online account to display your discounted pricing

(2S,4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid benzylamine salt, 95%, 98% ee, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: fisher scientific acros  362012500

 View more versions of this product

Catalog No. AC362012500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94% (HPLC)
Formula Weight 352.43
Physical Form Crystalline Powder
Quantity 250mg
Color White
Chemical Name or Material (2S, 4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid benzylamine salt
CAS 441044-14-0
Molecular Formula C11H19NO5·C7H9N
Assay Percent Range 95%
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% (HPLC) min.
Packaging Glass bottle

Specifications

Chemical Name or Material (2S, 4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid benzylamine salt
Assay ≥94% (HPLC)
Assay Percent Range 95%
Molecular Formula C11H19NO5·C7H9N
Formula Weight 352.43