Please login to your online account to display your discounted pricing

(2S,4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid benzylamine salt, 95%, 98% ee, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (2S, 4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid benzylamine salt
Assay ≥94% (HPLC)
Assay Percent Range 95%
Molecular Formula C11H19NO5·C7H9N
Formula Weight 352.43
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362012500

  SDS
fisher scientific acros
362012500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC362015000

  SDS
fisher scientific acros
362015000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC362010025

  SDS
thermo fisher scientific
AC362010025
Glass bottle 2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94% (HPLC)
Physical Form Crystalline Powder
Formula Weight 352.43
Quantity 250mg
Color White
Chemical Name or Material (2S, 4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid benzylamine salt
Molecular Formula C11H19NO5·C7H9N
CAS 441044-14-0
Assay Percent Range 95%
Enantiomeric Excess 97.5% (HPLC) min.
Infrared Spectrum Authentic
Packaging Glass bottle