missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic Acid 96.0+%, TCI America™
Supplier: TCI America B3177200MG
Specifications
| (2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic Acid | |
| 116°C | |
| C10H16FNO4 | |
| 200 mg | |
| YGWZXQOYEBWUTH-RQJHMYQMSA-N | |
| (2S,4R)-4-fluoro-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid | |
| 12043048 | |
| ≥96.0% (GC,T) |
| 203866-14-2 | |
| White-Yellow | |
| MFCD06796085 | |
| n-boc-trans-4-fluoro-l-proline, 2s,4r-1-tert-butoxycarbonyl-4-fluoropyrrolidine-2-carboxylic acid, boc-trans-4-fluoro-l-proline, n-tert-butoxycarbonyl-2s,4r-4-fluoroproline, 2s,4r-1-boc-4-fluoro-2-pyrrolidinecarboxylic acid, boc-trans-4-fluoro-pro-oh, 2s,4r-1-tert-butoxycarbonyl-4-fluoro-2-pyrrolidinecarboxylic acid, 2s,4r-1-tert-butoxy carbonyl-4-fluoropyrrolidine-2-carboxylic acid, trans-4-fluoro-l-proline, n-boc protected, 2s,4r-1-tert-butoxycarbonyl-4-fluoropyrrolidine-2-carboyxlic acid | |
| CC(C)(C)OC(=O)N1CC(CC1C(=O)O)F | |
| 233.239 | |
| 233.24 | |
| Crystalline Powder |
Chemical Identifiers
| 203866-14-2 | |
| 233.239 | |
| YGWZXQOYEBWUTH-RQJHMYQMSA-N | |
| 12043048 | |
| CC(C)(C)OC(=O)N1CC(CC1C(=O)O)F |
| C10H16FNO4 | |
| MFCD06796085 | |
| n-boc-trans-4-fluoro-l-proline, 2s,4r-1-tert-butoxycarbonyl-4-fluoropyrrolidine-2-carboxylic acid, boc-trans-4-fluoro-l-proline, n-tert-butoxycarbonyl-2s,4r-4-fluoroproline, 2s,4r-1-boc-4-fluoro-2-pyrrolidinecarboxylic acid, boc-trans-4-fluoro-pro-oh, 2s,4r-1-tert-butoxycarbonyl-4-fluoro-2-pyrrolidinecarboxylic acid, 2s,4r-1-tert-butoxy carbonyl-4-fluoropyrrolidine-2-carboxylic acid, trans-4-fluoro-l-proline, n-boc protected, 2s,4r-1-tert-butoxycarbonyl-4-fluoropyrrolidine-2-carboyxlic acid | |
| (2S,4R)-4-fluoro-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
Safety and Handling
TSCA : No