Please login to your online account to display your discounted pricing

(2S)-4-(1,3-Dioxoisoindolin-2-yl)-2-hydroxybutanoic acid, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (2S)-4-(1, 3-Dioxoisoindolin-2-yl)-2-hydroxybutanoic acid
Name Note 96%
CAS 48172-10-7
CAS Min % 95.0
CAS Max % 100.0
Assay ≥96%
Assay Percent Range 96%
Molecular Formula C12 H11 N O5
Formula Weight 249.22
MDL Number MFCD00192213
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC342450050

  SDS
thermo fisher scientific
342450050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC342450250

  SDS
thermo fisher scientific
342450250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96%
Physical Form Crystalline Powder
Formula Weight 249.22
Quantity 5g
Color White
TSCA TSCA
Melting Point 156° to 159°C
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material (2S)-4-(1, 3-Dioxoisoindolin-2-yl)-2-hydroxybutanoic acid
Packaging Glass bottle
Specific Rotation +6° to +7° (20°C, 589nm) (c=1, ethanol)
Name Note 96%
Assay Percent Range 96%
Molecular Formula C12 H11 N O5
MDL Number MFCD00192213
CAS 48172-10-7
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.