Please login to your online account to display your discounted pricing

Alfa Aesar™ (2S,3S)-(-)-Bis(diphenylphosphino)butane, 98%

$519.84 - $519.84

Chemical Identifiers

Chemical Name or Material (2S,3S)-(-)-Bis(diphenylphosphino)butane
CAS 64896-28-2
Molecular Formula C28H28P2
Formula Weight 426.48
MDL Number MFCD00009836
Beilstein 4268869
Synonym (S,S)-Chiraphos
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2758603

  SDS
alfa aesar™
H2758603
1g Each for $519.84
Description & Specifications

Specifications

Formula Weight 426.48
Quantity 1g
Health Hazard 1 Warning
Chemical Name or Material (2S,3S)-(-)-Bis(diphenylphosphino)butane
Beilstein 4268869
Synonym (S,S)-Chiraphos
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
MDL Number MFCD00009836
Molecular Formula C28H28P2
Melting Point 107° to 109°C
CAS 64896-28-2
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.