missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2S,3S)-3-(tert-Butoxycarbonylamino)-1,2-epoxy-4-phenylbutane, TCI America™

Manufacturer:  TCI America E089325G

 View more versions of this product

Catalog No. E089325G



Specifications

Specifications

(2S,3S)-3-(tert-Butoxycarbonylamino)-1,2-epoxy-4-phenylbutane
127°C
98737-29-2
MFCD02258997
2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, 2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, 2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate, 2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane, ksc523o6b
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2
263.337
263.34
≥98.0% (GC)
White-Yellow
25g
C15H21NO3
3077
NVPOUMXZERMIJK-QWHCGFSZSA-N
tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate
9903372
Crystal-Powder at 20°C

Chemical Identifiers

98737-29-2
263.337
NVPOUMXZERMIJK-QWHCGFSZSA-N
9903372
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2
C15H21NO3
MFCD02258997
2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, 2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, 2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate, 2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane, ksc523o6b
tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6533

TSCA : No

Documents