Learn More
(2S,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 395020050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(2S, 3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane | |
97.5 | |
97.5% min. (HPLC) | |
C15H21NO3 | |
5g | |
Solubility in water: <0.1% (20°C) | |
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2 | |
263.34 | |
263.34 |
98737-29-2 | |
100.0 | |
Glass bottle | |
MFCD02258997 | |
2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, 2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, 2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate, 2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane, ksc523o6b | |
NVPOUMXZERMIJK-QWHCGFSZSA-N | |
tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate | |
9903372 | |
98% |
Chemical Identifiers
98737-29-2 | |
263.34 | |
NVPOUMXZERMIJK-QWHCGFSZSA-N | |
9903372 | |
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2 |
C15H21NO3 | |
MFCD02258997 | |
2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, 2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, 2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate, 2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane, ksc523o6b | |
tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only