missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(2S,3S)-3-Methylpyrrolidine-2-carboxylic acid, 97%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific 290092500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(2S, 3S)-3-Methylpyrrolidine-2-carboxylic acid | |
White to Beige | |
Authentic | |
Glass bottle | |
250mg | |
CC1CCNC1C(=O)O | |
(2S,3S)-3-methylpyrrolidine-2-carboxylic acid | |
129.16 | |
129.16 | |
97% |
10512-89-7 | |
97% | |
C6H11NO2 | |
2s,3s-3-methylpyrrolidine-2-carboxylic acid, 2s,3s-3-methylpyrrolidine-2-carboxylicacid, 3beta-methyl-l-proline, 3s-3-methyl-l-proline, h-l-trans-pro 3-me-oh, l-proline, 3-methyl-, 3s, l-trans-3-methylproline, 2s,3s-3-methyl-pyrroline-2-carboxylic acid, trans-3-methyl-l-proline, trans-3-methylproline | |
CNPSFBUUYIVHAP-WHFBIAKZSA-N | |
− 28.00 (25.00°C c=0.7,water) | |
− 28.00 | |
2724879 | |
Crystalline Powder |
Chemical Identifiers
10512-89-7 | |
129.16 | |
2s,3s-3-methylpyrrolidine-2-carboxylic acid, 2s,3s-3-methylpyrrolidine-2-carboxylicacid, 3beta-methyl-l-proline, 3s-3-methyl-l-proline, h-l-trans-pro 3-me-oh, l-proline, 3-methyl-, 3s, l-trans-3-methylproline, 2s,3s-3-methyl-pyrroline-2-carboxylic acid, trans-3-methyl-l-proline, trans-3-methylproline | |
(2S,3S)-3-methylpyrrolidine-2-carboxylic acid |
C6H11NO2 | |
CNPSFBUUYIVHAP-WHFBIAKZSA-N | |
2724879 | |
CC1CCNC1C(=O)O |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only