missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (2S,3S)-3-(Boc-amino)-1,2-epoxy-4-phenylbutane, 98%

Manufacturer:  Alfa Aesar™ H5600106

 View more versions of this product

Catalog No. AAH5600106



Description

Description

Applications
It function as a building block employed in the synthesis of HIV-1 protease inhibitors and is also employed in synthesizing (hydroxyethyl)urea peptidomimetrics and arylsulfonamides possessing anti-HIV activity. It is a Enantiomer S. Atazanavir intermediate.

Solubility
Insoluble in water. Soluble in Chloroform, Dichloromethane and Ethyl Acetate.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

(2S,3S)-3-(Boc-amino)-1,2-epoxy-4-phenylbutane
125°C to 127°C
98737-29-2
MFCD02258997
2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane
NVPOUMXZERMIJK-QWHCGFSZSA-N
tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate
9903372
98%
−71° (c=0.6 in Methanol)
5g
C15H21NO3
UN3077
Insoluble in water. Soluble in Chloroform,Dichloromethane and Ethyl Acetate.
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2
263.337
263.34

Chemical Identifiers

98737-29-2
263.337
NVPOUMXZERMIJK-QWHCGFSZSA-N
9903372
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2
C15H21NO3
MFCD02258997
2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane
tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 425-420-5

TSCA : No

Recommended Storage : Ambient temperatures

Documents