missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(2S,3S)-(+)-2,3-Butanediol, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 247735000
Description
Chiral derivatization reagentSpecifications
(2S, 3S)-(+)-2, 3-Butanediol | |
85°C | |
Glass bottle | |
0.98 | |
179.0°C to 182.0°C | |
500mg | |
19132-06-0 | |
100.0 | |
C4H10O2 | |
MFCD00063648 | |
01,82 | |
2,3-butanediol, +, 2,3-butanediol, s-r*,r*, 2s,3s-+-2,3-butanediol, 2s,3s-2,3-butanediol, 2s,3s-butane-2,3-diol, s,s-+-2,3-butanediol, s,s-2,3-butanediol, s,s-2,3-butylene glycol, s,s-butane-2,3-diol, unii-7e9uxg71s1 | |
OWBTYPJTUOEWEK-IMJSIDKUSA-N | |
(2S,3S)-butane-2,3-diol | |
439888 | |
90.12 | |
99% |
0.9800g/mL | |
Authentic | |
1.4309 to 1.4329 | |
+ 13.00 | |
Yellow | |
99% | |
98.5 | |
98.5% min. (GC) | |
CH3CH(OH)CH(OH)CH3 | |
01, 2183 | |
15, 1571 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and diisopropyl ether | |
CC(C(C)O)O | |
90.12 | |
CHEBI:16812 | |
Oily Liquid |
Chemical Identifiers
19132-06-0 | |
90.12 | |
OWBTYPJTUOEWEK-IMJSIDKUSA-N | |
439888 | |
(2S,3S)-butane-2,3-diol |
C4H10O2 | |
MFCD00063648 | |
2,3-butanediol, +, 2,3-butanediol, s-r*,r*, 2s,3s-+-2,3-butanediol, 2s,3s-2,3-butanediol, 2s,3s-butane-2,3-diol, s,s-+-2,3-butanediol, s,s-2,3-butanediol, s,s-2,3-butylene glycol, s,s-butane-2,3-diol, unii-7e9uxg71s1 | |
CHEBI:16812 | |
CC(C(C)O)O |
Safety and Handling
AIR SENSITIVE
HYGROSCOPIC
TSCA : TSCA