missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2S,3R)-(+)-2-Amino-3-hydroxy-4-methylpentanoic acid, 99%, 99+% ee, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 290072500

 View more versions of this product

Catalog No. AC290072500



Specifications

Specifications

(2S,3R)-(+)-2-Amino-3-hydroxy-4-methylpentanoicacid
Authentic
+ 18.50 (25.00°C c=1,6N HCl)
White
250mg
99%
(CH3)2CHCH(OH)CH(NH2)CO2H
ZAYJDMWJYCTABM-CRCLSJGQSA-N
(2S,3R)-2-amino-3-hydroxy-4-methylpentanoic acid
6994743
Fluffy Powder
> 99.0% (GC)
Glass bottle
+18.4° to +20.4° (25°C, 589nm) (c=2, 6N HCl)
211°C to 215°C
10148-71-7
C6H13NO3
2s*,3r*-2-amino-3-hydroxy-4-methyl-pentanoic acid, 2s,3r-, 2s,3r-+-2-amino-3-hydroxy-4-methylpentanoic acid, 2s,3r-2-amino-3-hydroxy-4-methyl-pentanoic acid, 2s,3r-2-amino-3-hydroxy-4-methylpentanoic acid, 2s,3r-3-hydroxyleucine, 2s,3r-beta-hydroxyleucine, 3r-3-hydroxy-l-leucine, a-hydroxyleucine, l-leucine, 3-hydroxy-, 3r, l-threo-
CC(C)C(C(C(=O)O)N)O
147.174
147.17
≥98.5% (TLC)

Chemical Identifiers

10148-71-7
147.174
2s*,3r*-2-amino-3-hydroxy-4-methyl-pentanoic acid, 2s,3r-, 2s,3r-+-2-amino-3-hydroxy-4-methylpentanoic acid, 2s,3r-2-amino-3-hydroxy-4-methyl-pentanoic acid, 2s,3r-2-amino-3-hydroxy-4-methylpentanoic acid, 2s,3r-3-hydroxyleucine, 2s,3r-beta-hydroxyleucine, 3r-3-hydroxy-l-leucine, a-hydroxyleucine, l-leucine, 3-hydroxy-, 3r, l-threo-
(2S,3R)-2-amino-3-hydroxy-4-methylpentanoic acid
C6H13NO3
ZAYJDMWJYCTABM-CRCLSJGQSA-N
6994743
CC(C)C(C(C(=O)O)N)O
Documents