missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2S,3R)-(+)-2-Amino-3-hydroxy-4-methylpentanoic acid, 99%, 99+% ee, ACROS Organics™

$639.75 - $639.75

Chemical Identifiers

CAS 10148-71-7
Molecular Formula C6H13NO3
Molecular Weight (g/mol) 147.174
InChI Key ZAYJDMWJYCTABM-CRCLSJGQSA-N
Synonym 2s*,3r*-2-amino-3-hydroxy-4-methyl-pentanoic acid
PubChem CID 6994743
IUPAC Name (2S,3R)-2-amino-3-hydroxy-4-methylpentanoic acid
SMILES CC(C)C(C(C(=O)O)N)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC290072500 View Documents Thermo Fisher Scientific
290072500
250mg Glass bottle Each for $639.75
 
Specifications

Chemical Identifiers

10148-71-7
147.174
2s*,3r*-2-amino-3-hydroxy-4-methyl-pentanoic acid
(2S,3R)-2-amino-3-hydroxy-4-methylpentanoic acid
C6H13NO3
ZAYJDMWJYCTABM-CRCLSJGQSA-N
6994743
CC(C)C(C(C(=O)O)N)O

Specifications

> 99.0% (GC)
Glass bottle
+18.4° to +20.4° (25°C, 589nm) (c=2, 6N HCl)
211°C to 215°C
10148-71-7
C6H13NO3
2s*,3r*-2-amino-3-hydroxy-4-methyl-pentanoic acid
CC(C)C(C(C(=O)O)N)O
147.174
147.17
≥98.5% (TLC)
Authentic
+ 18.50 (25.00°C c=1,6N HCl)
White
250mg
99%
(CH3)2CHCH(OH)CH(NH2)CO2H
ZAYJDMWJYCTABM-CRCLSJGQSA-N
(2S,3R)-2-amino-3-hydroxy-4-methylpentanoic acid
6994743
Fluffy Powder
(2S, 3R)-(+)-2-Amino-3-hydroxy-4-methylpentanoic acid