missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2S,3R)-(+)-2-Amino-3-hydroxy-4-methylpentanoic acid, 99%, 99+% ee, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC290070010

 View more versions of this product

Catalog No. AC290070010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Fluffy Powder
≥98.5% (TLC)
White
+ 18.50 (25.00°C c=1,6N HCl)
> 99.0% (GC)
C6H13NO3
Glass bottle
+18.4° to +20.4° (25°C, 589nm) (c=2, 6N HCl)
(CH3)2CHCH(OH)CH(NH2)CO2H
147.17
1g
211° to 215°C
(2S, 3R)-(+)-2-Amino-3-hydroxy-4-methylpentanoic acid
10148-71-7
Authentic
99%
L(+)-threo-3-Hydroxyleucine

Chemical Identifiers

99%
(CH3)2CHCH(OH)CH(NH2)CO2H
L(+)-threo-3-Hydroxyleucine
≥98.5% (TLC)
C6H13NO3
147.17
(2S, 3R)-(+)-2-Amino-3-hydroxy-4-methylpentanoic acid