missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2R)-Bornane-10,2-sultam, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 294190010

 View more versions of this product

Catalog No. AC294190010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(2R)-Bornane-10, 2-sultam
Authentic
-33
181°C to 185°C
≥99% ee
99% min. (GC)
MFCD00066271
DPJYJNYYDJOJNO-SZBHIRRCSA-N
215.32
215.32
99+%
> 99%
Glass bottle
Colorless
1g
94594-90-8
C10H17NO2S
--10,2-camphorsultam;--exo-10,2-bornanesultam; 3as-3a, 5r,7r-10,10-dimethyl-3??-thia-4-azatricyclo 5.2.1.0 1 ,? decane-3,3-dione, a,6, a,7a, a-hexahydro-8,8-dimethyl-3h-3a,6-methano-2,1-benzisothiazole-2,2-dioxide; 1s,5r-10,10-dimethyl-3-thia-4-azatricyclo 5.2.1.01,5 decane 3,3-dioxide
CC1(C2CCC13CS(=O)(=O)NC3C2)C
54579826
Liquid

Chemical Identifiers

94594-90-8
215.32
DPJYJNYYDJOJNO-SZBHIRRCSA-N
54579826
C10H17NO2S
MFCD00066271
--10,2-camphorsultam;--exo-10,2-bornanesultam; 3as-3a, 5r,7r-10,10-dimethyl-3??-thia-4-azatricyclo 5.2.1.0 1 ,? decane-3,3-dione, a,6, a,7a, a-hexahydro-8,8-dimethyl-3h-3a,6-methano-2,1-benzisothiazole-2,2-dioxide; 1s,5r-10,10-dimethyl-3-thia-4-azatricyclo 5.2.1.01,5 decane 3,3-dioxide
CC1(C2CCC13CS(=O)(=O)NC3C2)C
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents