Please login to your online account to display your discounted pricing

(2R,4R)-APDC, Tocris

Highly selective group II agonist

$309.99 - $1,284.01

Chemical Identifiers

Chemical Name or Material (2R, 4R)-APDC
Assay Percent Range >98%
Molecular Formula C6H10N2O4
Formula Weight 174.16
Synonym (2R, 4R)-4-Aminopyrrolidine-2, 4-dicarboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

120850

 
r & d systems
120850
50mg Each for $1,284.01

120810

 
r & d systems
120810
10mg Each for $309.99
Description & Specifications

Specifications

Formula Weight 174.16
Quantity 50mg
Chemical Name or Material (2R, 4R)-APDC
Assay Percent Range >98%
Molecular Formula C6H10N2O4
CAS 169209-63-6
Recommended Storage Desiccate at Room Temperature
Synonym (2R, 4R)-4-Aminopyrrolidine-2, 4-dicarboxylate
Safety and Handling

Desiccate at Room Temperature