Please login to your online account to display your discounted pricing

(2R,3S)-1-Carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt, 85%, Tech., ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC371002500

 View more versions of this product

Catalog No. AC371002500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥80%
Purity Grade Technical Grade
Physical Form Crystalline Powder
Formula Weight 264.35
Quantity 250mg
Color Off-white to beige
CAS Min % 80.0
CAS Max % 100.0
Chemical Name or Material (2R,3S)-1-Carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt
Infrared Spectrum Authentic
Name Note tech., 85%
Assay Percent Range 85%
Packaging Glass bottle
MDL Number MFCD00142975
CAS 205639-93-6
Recommended Storage Freezer -20°C
Molecular Formula C12H17KO4
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (2R,3S)-1-Carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt
Name Note tech., 85%
CAS 205639-93-6
CAS Min % 80.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥80%
Assay Percent Range 85%
Molecular Formula C12H17KO4
Formula Weight 264.35
MDL Number MFCD00142975
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

Freezer -20°C

SDS & Safety Documents