missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2R,3S)-1-Carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt, 85%, Tech., Thermo Scientific™

Chemical Identifiers

CAS 205639-93-6
Molecular Formula C12H17KO4
Molecular Weight (g/mol) 264.35
MDL Number MFCD00142975
InChI Key BBLPZPQHEWQLGM-VZXYPILPSA-M
Synonym potassium 5s,6r-5,6-dihydroxy-4-pentylcyclohexa-1,3-dienecarboxylate, 2r,3s-1-carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt
PubChem CID 23666349
IUPAC Name potassium;(5S,6R)-5,6-dihydroxy-4-pentylcyclohexa-1,3-diene-1-carboxylate
SMILES CCCCCC1=CC=C(C(C1O)O)C(=O)[O-].[K+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC371002500
SDS View Documents Promotion Details
Thermo Scientific Chemicals
371002500
250mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

205639-93-6
264.35
BBLPZPQHEWQLGM-VZXYPILPSA-M
23666349
CCCCCC1=CC=C(C(C1O)O)C(=O)[O-].[K+]
C12H17KO4
MFCD00142975
potassium 5s,6r-5,6-dihydroxy-4-pentylcyclohexa-1,3-dienecarboxylate, 2r,3s-1-carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt
potassium;(5S,6R)-5,6-dihydroxy-4-pentylcyclohexa-1,3-diene-1-carboxylate

Specifications

205639-93-6
100.0
Authentic
MFCD00142975
250mg
BBLPZPQHEWQLGM-VZXYPILPSA-M
potassium;(5S,6R)-5,6-dihydroxy-4-pentylcyclohexa-1,3-diene-1-carboxylate
23666349
Crystalline Powder
Technical
80.0
80% min. (HPLC)
C12H17KO4
Glass bottle
potassium 5s,6r-5,6-dihydroxy-4-pentylcyclohexa-1,3-dienecarboxylate, 2r,3s-1-carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt
CCCCCC1=CC=C(C(C1O)O)C(=O)[O-].[K+]
264.35
264.35
85%
(2R,3S)-1-Carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt, tech., 85%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Freezer -20°C

SDS

RUO – Research Use Only