Please login to your online account to display your discounted pricing

(2R,3S)-1-Carboxy-4-ethyl-2,3-dihydroxy-cyclohexa-4,6-diene, sodium salt, 85%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  271802500

 View more versions of this product

Catalog No. AC271802500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥80%
Physical Form Crystalline Powder
Formula Weight 206.17
Quantity 250mg
Color Off-white to light beige
TSCA TSCA
CAS Min % 80.0
CAS Max % 100.0
Chemical Name or Material (2R,3S)-1-Carboxy-4-ethyl-2,3-dihydroxy-cyclohexa-4,6-diene, sodium salt
MDL Number MFCD00142973
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 85%
CAS 205652-53-5
Name Note 85%
Molecular Formula C9 H11 Na O4
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (2R,3S)-1-Carboxy-4-ethyl-2,3-dihydroxy-cyclohexa-4,6-diene, sodium salt
Name Note 85%
CAS 205652-53-5
CAS Min % 80.0
CAS Max % 100.0
Assay ≥80%
Assay Percent Range 85%
Molecular Formula C9 H11 Na O4
Formula Weight 206.17
MDL Number MFCD00142973
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.

SDS & Safety Documents