Please login to your online account to display your discounted pricing

(2R,3S)-1-Carboxy-4-ethyl-2,3-dihydroxy-cyclohexa-4,6-diene, sodium salt, 85%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  271800010

 View more versions of this product

Catalog No. AC271800010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥80%
Formula Weight 206.17
Physical Form Crystalline Powder
Quantity 1g
Color Off-white to light beige
TSCA TSCA
CAS Max % 100.0
CAS Min % 80.0
Name Note 85%
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C9 H11 Na O4
MDL Number MFCD00142973
Chemical Name or Material (2R,3S)-1-Carboxy-4-ethyl-2,3-dihydroxy-cyclohexa-4,6-diene, sodium salt
CAS 205652-53-5
Assay Percent Range 85%
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.

Specifications

Chemical Name or Material (2R,3S)-1-Carboxy-4-ethyl-2,3-dihydroxy-cyclohexa-4,6-diene, sodium salt
Name Note 85%
CAS 205652-53-5
CAS Min % 80.0
CAS Max % 100.0
Assay ≥80%
Assay Percent Range 85%
Molecular Formula C9 H11 Na O4
Formula Weight 206.17
MDL Number MFCD00142973
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.

SDS & Safety Documents