Please login to your online account to display your discounted pricing

(2R,3S)-1-Carboxy-4-ethyl-2,3-dihydroxy-cyclohexa-4,6-diene, sodium salt, 85%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (2R,3S)-1-Carboxy-4-ethyl-2,3-dihydroxy-cyclohexa-4,6-diene, sodium salt
Name Note 85%
CAS 205652-53-5
CAS Min % 80.0
CAS Max % 100.0
Assay ≥80%
Assay Percent Range 85%
Molecular Formula C9 H11 Na O4
Formula Weight 206.17
MDL Number MFCD00142973
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC271800010

  SDS
thermo fisher scientific
271800010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC271802500

  SDS
thermo fisher scientific
271802500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥80%
Formula Weight 206.17
Physical Form Crystalline Powder
Quantity 1g
Color Off-white to light beige
TSCA TSCA
CAS Max % 100.0
CAS Min % 80.0
Name Note 85%
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C9 H11 Na O4
MDL Number MFCD00142973
Chemical Name or Material (2R,3S)-1-Carboxy-4-ethyl-2,3-dihydroxy-cyclohexa-4,6-diene, sodium salt
CAS 205652-53-5
Assay Percent Range 85%
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.