missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2R,3R)-(-)-2,3-Butanediol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0658603

 View more versions of this product

Catalog No. AAL0658603



Description

Description

(2R,3R)-(-)-2,3-Butanediol is used as a chiral auxiliary, chiral ligand and building block. Cyclocondenses with ketones for (13)C NMR determination of optical purity. It is used in the resolution of carbonyl compounds in gas chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(2R,3R)-(-)-2,3-Butanediol is used as a chiral auxiliary, chiral ligand and building block. Cyclocondenses with ketones for (13)C NMR determination of optical purity. It is used in the resolution of carbonyl compounds in gas chromatography.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store away from oxidizing agents. Incompatible with oxidizing agents, Acid chlorides, Acid anhydrides, Chloroformates, Reducing agents. Air sensitive.
Specifications

Specifications

(2R,3R)-(-)-2,3-Butanediol
16°C
179°C to 180°C
C4H10O2
MFCD00064267
4290593
14,1568
Fully miscible in water.
−13° (Neat)
(2R,3R)-butane-2,3-diol
225936
90.12
24347-58-8
0.988
85°C (185°F)
1.4315
1 g
Hygroscopic
2r,3r-butane-2,3-diol, r,r-2,3-butanediol, 2r,3r---2,3-butanediol, r,r-butane-2,3-diol, 2r,3r-2,3-butanediol, r,r-2,3-butylene glycol, r,r---butane-2,3-diol, r,r---2,3-butylene glycol, unii-or02b2286a, r,r---2,3-butanediol
OWBTYPJTUOEWEK-UHFFFAOYNA-N
CC(O)C(C)O
90.12
CHEBI:16982
98%

Chemical Identifiers

24347-58-8
90.12
OWBTYPJTUOEWEK-UHFFFAOYNA-N
225936
(2R,3R)-butane-2,3-diol
C4H10O2
MFCD00064267
2r,3r-butane-2,3-diol, r,r-2,3-butanediol, 2r,3r---2,3-butanediol, r,r-butane-2,3-diol, 2r,3r-2,3-butanediol, r,r-2,3-butylene glycol, r,r---butane-2,3-diol, r,r---2,3-butylene glycol, unii-or02b2286a, r,r---2,3-butanediol
CHEBI:16982
CC(O)C(C)O
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 246-186-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only