Please login to your online account to display your discounted pricing

Alfa Aesar™ 2H-Pyrido[3,2-b]-1,4-oxazin-3(4H)-one, 98%

$32.39 - $32.39

Chemical Identifiers

Chemical Name or Material 2H-Pyrido[3,2-b]-1,4-oxazin-3(4H)-one
CAS 20348-09-8
Molecular Formula C7H6N2O2
Formula Weight 150.14
MDL Number MFCD00006697
Beilstein 1074010
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0847303

  SDS
alfa aesar™
L0847303
1g Each for $32.39
Description & Specifications

Specifications

Formula Weight 150.14
Chemical Name or Material 2H-Pyrido[3,2-b]-1,4-oxazin-3(4H)-one
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
MDL Number MFCD00006697
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 1074010
Health Hazard 1 Warning
Quantity 1g
Melting Point 204° to 208°C
CAS 20348-09-8
Molecular Formula C7H6N2O2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.