missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2H-Pyran-2-one, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 376810050

 View more versions of this product

Catalog No. AC376810050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2H-Pyran-2-one
96.0
1.2100g/mL
94°C
96% min. (GC)
C5H4O2
MFCD00006639
1.21
ZPSJGADGUYYRKE-UHFFFAOYSA-N
pyran-2-one
68154
96.09
504-31-4
100.0
104°C (30.0mmHg)
Authentic
Glass bottle
1.5280 to 1.531
5g
2h-pyran-2-one, alpha-pyrone, 2-pyranone, 2-pyrone, coumalin, pyrone, .alpha.-pyrone, unii-8ww45i202v, 2-oxo-2h-pyran, oxidopyrylium
C1=CC(=O)OC=C1
96.09
CHEBI:37965
97%

Chemical Identifiers

504-31-4
96.09
ZPSJGADGUYYRKE-UHFFFAOYSA-N
68154
pyran-2-one
C5H4O2
MFCD00006639
2h-pyran-2-one, alpha-pyrone, 2-pyranone, 2-pyrone, coumalin, pyrone, .alpha.-pyrone, unii-8ww45i202v, 2-oxo-2h-pyran, oxidopyrylium
CHEBI:37965
C1=CC(=O)OC=C1
Safety and Handling

Safety and Handling

EINECSNumber : 207-990-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only