Please login to your online account to display your discounted pricing

2H-Pyran-2-one, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC376810010

 View more versions of this product

Catalog No. AC376810010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Formula Weight 96.09
Quantity 1g
Color Clear yellow to brown
Density 1.2100g/mL
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
CAS 504-31-4
Assay Percent Range 97%
Molecular Formula C5H4O2
MDL Number MFCD00006639
Refractive Index 1.5280 to 1.5310 (20°C, 589nm)
Packaging Glass bottle
Synonym α-Pyrone
Chemical Name or Material 2H-Pyran-2-one

Specifications

Chemical Name or Material 2H-Pyran-2-one
CAS 504-31-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H4O2
Formula Weight 96.09
MDL Number MFCD00006639
Synonym α-Pyrone