Please login to your online account to display your discounted pricing

Alfa Aesar™ 2H-Perfluoro-5-methyl-3,6-dioxanonane, 97%


Quantity 25g

Manufacturer: alfa aesar™  L1695414

 View more versions of this product

Catalog No. AAL1695414

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 452.07
Chemical Name or Material 2H-Perfluoro-5-methyl-3,6-dioxanonane
CAS 3330-14-1
Molecular Formula C8HF17O2
Synonym Fluoroether E2; 2H-Heptadecafluoro-5-methyl-3,6-dioxanonane
Assay 97%
TSCA TSCA
Melting Point −122°C
Boiling Point 103° to 105°C
Quantity 25g
MDL Number MFCD00054714

Specifications

Chemical Name or Material 2H-Perfluoro-5-methyl-3,6-dioxanonane
CAS 3330-14-1
Molecular Formula C8HF17O2
Formula Weight 452.07
MDL Number MFCD00054714
Synonym Fluoroether E2; 2H-Heptadecafluoro-5-methyl-3,6-dioxanonane