Please login to your online account to display your discounted pricing

Alfa Aesar™ 2H-Perfluoro-5-methyl-3,6-dioxanonane, 97%

$201.65 - $863.54

Chemical Identifiers

Chemical Name or Material 2H-Perfluoro-5-methyl-3,6-dioxanonane
CAS 3330-14-1
Molecular Formula C8HF17O2
Formula Weight 452.07
MDL Number MFCD00054714
Synonym Fluoroether E2; 2H-Heptadecafluoro-5-methyl-3,6-dioxanonane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1695406

 
alfa aesar™
L1695406
5g Each for $201.65

AAL1695414

 
alfa aesar™
L1695414
25g Each for $863.54
Description & Specifications

Specifications

Formula Weight 452.07
Chemical Name or Material 2H-Perfluoro-5-methyl-3,6-dioxanonane
TSCA TSCA
Assay 97%
Molecular Formula C8HF17O2
MDL Number MFCD00054714
CAS 3330-14-1
Synonym Fluoroether E2; 2H-Heptadecafluoro-5-methyl-3,6-dioxanonane
Quantity 5g
Boiling Point 103° to 105°C
Melting Point −122°C