missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2H-Perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxapentadecane

$97.01 - $371.65

Chemical Identifiers

CAS 26738-51-2
Molecular Formula C14HF29O4
Molecular Weight (g/mol) 784.112
MDL Number MFCD00054717
InChI Key NOCLWIPRQIWFMR-UHFFFAOYSA-N
Synonym 2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-icosafluoro-5,8,11-tris trifluoromethyl-3,6,9,12-tetraoxapentadecane, 3,6,9,12-tetraoxapentadecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-eicosafluoro-5,8,11-tris trifluoromethyl, freon e 4, freon e-4, 2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxapentadecane, 2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane, 1,1,1,2,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy-3-1,2,2,2-tetrafluoroethoxy propane
PubChem CID 117327
IUPAC Name 1,1,1,2,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy]-3-(1,2,2,2-tetrafluoroethoxy)propane
SMILES C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F)(F)F)F)(F)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1695306 View Documents Alfa Aesar™
L1695306
5g Each for $97.01
 
AAL1695314 View Documents Alfa Aesar™
L1695314
25g Each for $371.65
 
Specifications

Chemical Identifiers

26738-51-2
784.112
NOCLWIPRQIWFMR-UHFFFAOYSA-N
117327
C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F)(F)F)F)(F)F)F
C14HF29O4
MFCD00054717
2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-icosafluoro-5,8,11-tris trifluoromethyl-3,6,9,12-tetraoxapentadecane, 3,6,9,12-tetraoxapentadecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-eicosafluoro-5,8,11-tris trifluoromethyl, freon e 4, freon e-4, 2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxapentadecane, 2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane, 1,1,1,2,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy-3-1,2,2,2-tetrafluoroethoxy propane
1,1,1,2,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy]-3-(1,2,2,2-tetrafluoroethoxy)propane

Specifications

192°C to 195°C
5g
C14HF29O4
2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-icosafluoro-5,8,11-tris trifluoromethyl-3,6,9,12-tetraoxapentadecane, 3,6,9,12-tetraoxapentadecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-eicosafluoro-5,8,11-tris trifluoromethyl, freon e 4, freon e-4, 2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxapentadecane, 2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane, 1,1,1,2,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy-3-1,2,2,2-tetrafluoroethoxy propane
C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F)(F)F)F)(F)F)F
784.112
784.12
-115°C
26738-51-2
MFCD00054717
NOCLWIPRQIWFMR-UHFFFAOYSA-N
1,1,1,2,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy]-3-(1,2,2,2-tetrafluoroethoxy)propane
117327
2H-Perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxapentadecane
Safety and Handling

Safety and Handling

TSCA : TSCA