Please login to your online account to display your discounted pricing

Alfa Aesar™ 2H-Perfluoro-5,8,11,14-tetramethyl-3,6,9,12,15-pentaoxaoctadecane

$85.43 - $326.97

Chemical Identifiers

Chemical Name or Material 2H-Perfluoro-5,8,11,14-tetramethyl-3,6,9,12,15-pentaoxaoctadecane
CAS 37486-69-4
Molecular Formula C17HF35O5
Formula Weight 950.14
MDL Number MFCD00054716
Synonym Fluoroether E5
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1695206

  SDS
alfa aesar™
L1695206
5g Each for $85.43

AAL1695214

  SDS
alfa aesar™
L1695214
25g Each for $326.97
Description & Specifications

Specifications

Formula Weight 950.14
Chemical Name or Material 2H-Perfluoro-5,8,11,14-tetramethyl-3,6,9,12,15-pentaoxaoctadecane
CAS 37486-69-4
Molecular Formula C17HF35O5
Synonym Fluoroether E5
Quantity 5g
TSCA TSCA
Melting Point −95°C
Boiling Point 221° to 225°C
MDL Number MFCD00054716