Please login to your online account to display your discounted pricing

Alfa Aesar™ 2H-Perfluoro-5,8,11,14,17-pentamethyl-3,6,9,12,15,18-hexaoxaheneicosane

$288.34 - $288.34

Chemical Identifiers

Chemical Name or Material 2H-Perfluoro-5,8,11,14,17-pentamethyl-3,6,9,12,15,18-hexaoxaheneicosane
CAS 55154-18-2
Molecular Formula C20HF41O6
Formula Weight 1116.16g/mol
MDL Number MFCD06248658
Synonym Fluoroether E6
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1695114

 
alfa aesar™
L16951-14
25g Each for $288.34
Description & Specifications

Specifications

Boiling Point 108° to 110°C (1mmHg)
Melting Point −72°C
Quantity 25g
Chemical Name or Material 2H-Perfluoro-5,8,11,14,17-pentamethyl-3,6,9,12,15,18-hexaoxaheneicosane
CAS 55154-18-2
Molecular Formula C20HF41O6
Formula Weight 1116.16g/mol
MDL Number MFCD06248658
Synonym Fluoroether E6
TSCA TSCA