missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2H-Chromene-3-carboxylic acid, 90+%

Chemical Identifiers

Assay Percent Range 90%
Molecular Formula C10H8O3
Formula Weight 176.17
Chemical Name or Material 2H-Chromene-3-carboxylic acid
Assay 90%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC24101DA SDS Fisher Scientific Company
CC24101DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC24101EA Fisher Scientific Company
CC24101EA
Amber Glass Bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC24101CB SDS Fisher Scientific Company
CC24101CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

90%
176.17
90%
C10H8O3
2H-Chromene-3-carboxylic acid

Specifications

176.17
Amber Glass Bottle
C10H8O3
2H-Chromene-3-carboxylic acid
1g
90%
22649-28-1
90%