Please login to your online account to display your discounted pricing

2H-Chromene-3-carboxylic acid, 90+%


Quantity 250mg

Manufacturer: fisher scientific company  CC24101CB

 View more versions of this product

Catalog No. CC24101CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 176.17
Quantity 250mg
Assay 90%
Chemical Name or Material 2H-Chromene-3-carboxylic acid
Assay Percent Range 90%
Molecular Formula C10H8O3
CAS 22649-28-1

Specifications

Chemical Name or Material 2H-Chromene-3-carboxylic acid
Assay 90%
Assay Percent Range 90%
Molecular Formula C10H8O3
Formula Weight 176.17