Please login to your online account to display your discounted pricing

2H-Chromene-3-carbaldehyde, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2H-Chromene-3-carbaldehyde
CAS 51593-69-2
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H8O2
Formula Weight 160.17
MDL Number MFCD01662414
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC24104EA

 
fisher scientific company
CC24104EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC24104DA

 
fisher scientific company
CC24104DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 160.17
Quantity 10g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 2H-Chromene-3-carbaldehyde
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD01662414
CAS 51593-69-2
CAS Min % 96.5
Molecular Formula C10H8O2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.