Please login to your online account to display your discounted pricing

Alfa Aesar™ 2H,3H-Perfluoropentane, tech. 90%

$53.73 - $190.86

Chemical Identifiers

Chemical Name or Material 2H,3H-Perfluoropentane
CAS 138495-42-8
Purity Grade Technical Grade
Molecular Formula C5H2F10
Linear Formula CF3CHFCHFCF2CF3
Formula Weight 252.05
MDL Number MFCD00798129
Synonym 1,1,1,2,3,4,4,5,5,5-Decafluoropentane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2163306

 
alfa aesar™
B2163306
5g Each for $53.73

AAB2163314

 
alfa aesar™
B2163314
25g Each for $190.86
Description & Specifications

Specifications

Formula Weight 252.05
Chemical Name or Material 2H,3H-Perfluoropentane
TSCA TSCA
Purity Grade Technical Grade
Assay 90%
Molecular Formula C5H2F10
Linear Formula CF3CHFCHFCF2CF3
MDL Number MFCD00798129
CAS 138495-42-8
Synonym 1,1,1,2,3,4,4,5,5,5-Decafluoropentane
Boiling Point 54°C
Melting Point −80°C
Quantity 5g

Applications
Used as a zero ozone-depletion alternative to hydro chlorofluorocarbons.

Solubility
Soluble in water (140 mg/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.