Please login to your online account to display your discounted pricing

2H-1,4-Benzoxazin-3(4H)-one, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2H-1, 4-Benzoxazin-3(4H)-one
CAS 5466-88-6
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min. (HPLC)
Assay Percent Range 99%
Molecular Formula C8H7NO2
Formula Weight 149.15g/mol
MDL Number MFCD00158536
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC372850250

  SDS
thermo fisher scientific
372850250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color White to yellow to tan
Melting Point 172° to 176°C
Quantity 25g
Chemical Name or Material 2H-1, 4-Benzoxazin-3(4H)-one
CAS 5466-88-6
CAS Min % 98.5
CAS Max % 100.0
Physical Form White to yellow to tan flakes or powder
Assay 98.5% min. (HPLC)
Assay Percent Range 99%
Molecular Formula C8H7NO2
Formula Weight 149.15g/mol
MDL Number MFCD00158536
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.