Please login to your online account to display your discounted pricing

2H-1,4-Benzoxazin-3(4H)-one, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC372850250

 View more versions of this product

Catalog No. AC372850250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (HPLC)
Physical Form Flakes or Powder
Formula Weight 149.15
Quantity 25g
Color White to yellow to tan
Melting Point 172° to 176°C
CAS Max % 100.0
Molecular Formula C8H7NO2
MDL Number MFCD00158536
CAS 5466-88-6
Packaging Glass bottle
CAS Min % 98.5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2H-1, 4-Benzoxazin-3(4H)-one
Assay Percent Range 99%
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2H-1, 4-Benzoxazin-3(4H)-one
CAS 5466-88-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (HPLC)
Assay Percent Range 99%
Molecular Formula C8H7NO2
Formula Weight 149.15
MDL Number MFCD00158536
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents