Please login to your online account to display your discounted pricing

Alfa Aesar™ 2F5 Epitope

For cellular and molecular biology applications

Specifications

Product Type 2F5 epitope
Molecular Weight 1616.83
Storage Requirements -30° to -10°C
Sequence Glu-Leu-Leu-Glu-Leu-Asp-Lys-Trp-Ala-Ser-Leu-Trp-Asn
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66524EXD

 
alfa aesar™
J66524EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66524MCR

 
alfa aesar™
J66524MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type 2F5 epitope
Molecular Weight 1616.83
Storage Requirements -30° to -10°C
Sequence Glu-Leu-Leu-Glu-Leu-Asp-Lys-Trp-Ala-Ser-Leu-Trp-Asn
Format Solid
Regulatory Status RUO
Purification Method HPLC