missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2E,4E)-5-Phenyl-2,4-pentadienoic Acid, TCI America™

Manufacturer:  TCI America P242225G

 View more versions of this product

Catalog No. P242225G



Specifications

Specifications

(2E,4E)-5-Phenyl-2,4-pentadienoic Acid
≥98.0% (GC,T)
MFCD00014018
FEIQOMCWGDNMHM-KBXRYBNXSA-N
(2E,4E)-5-phenylpenta-2,4-dienoic acid
1549512
Crystal-Powder at 20°C
White
28010-12-0
C11H10O2
(E,E)-Cinnamylideneacetic Acid
C1=CC=C(C=C1)C=CC=CC(=O)O
174.199
174.20
168°C
25g

Chemical Identifiers

28010-12-0
174.199
FEIQOMCWGDNMHM-KBXRYBNXSA-N
1549512
C1=CC=C(C=C1)C=CC=CC(=O)O
C11H10O2
MFCD00014018
(E,E)-Cinnamylideneacetic Acid
(2E,4E)-5-phenylpenta-2,4-dienoic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents