Thermo Scientific™ Thermo Scientific™ 2D Tank Liner, ASI™ 26 film
Single-use tank liners reduce cleaning steps common to re-usable tanks.
$514.26 - $1,042.40
Specifications
Sterilization Method Irradiated
Product Line HyPerforma™
Shipping Condition Room Temperature
View More Specs
Products 7
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
B100037I
View Documents
Thermo Scientific™
B100037I
200 L
Case of 20 Each for $532.78
B100038I
View Documents
Thermo Scientific™
B100038I
130 L
Case of 20 Each for $514.26
B100048I
View Documents
Thermo Scientific™
B100048I
50L
Case of 40 Each for $946.80
B100049I
View Documents
Thermo Scientific™
B100049I
340 L
Case of 35 Each for $1,042.40
B100050I
View Documents
Thermo Scientific™
B100050I
400 L
Case of 20 Each for $636.20
B100051I
View Documents
Thermo Scientific™
B100051I
560 L
Case of 20 Each for $822.31
PV3687 DFS Item
View Documents
Thermo Scientific™
LSPV3687
Each for $592.00
Description
Description
The Thermo Scientific™ 2D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
  • Minimizes bioburden due to construction under sterile cGMP conditions
  • Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
Specifications
Irradiated
HyPerforma™
Room Temperature
SDS