missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Thermo Scientific™ 2D Tank Liner, ASI™ 26 film
Single-use tank liners reduce cleaning steps common to re-usable tanks.
Supplier: Thermo Scientific™ B100051I
Description
The Thermo Scientific™ 2D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
- Minimizes bioburden due to construction under sterile cGMP conditions
- Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
560 L | |
Store at room temperature. | |
20 per case | |
Irradiated | |
HyPerforma™ | |
ASI 26 |
Polyethylene | |
78.7 cm x 124.5 cm (31in. x 49 in.) | |
SV30116 | |
Room Temperature | |
0 Ports | |
No Tubing |