Please login to your online account to display your discounted pricing

29-Series Pop Beads


Create models of DNA strand, DNA replication, RNA conversion, cell organelles, ribosomes, mitochondria and a nucleus.

Manufacturer: fisher science education  29-2205

 View more versions of this product

Catalog No. S05939

  • / Each
 

Description & Specifications

Specifications

Color Yellow
Type 2 Way Pop Bead
Diameter (English) 0.5 in.
Diameter (Metric) 1.3cm
For Use With (Application) Create model of DNA strand, DNA replication, RNA conversion, cell organelles, ribosomes, mitochondria and nucleus