missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ 25mm Glass Microanalysis Vacuum Filter Holder and Support

Vacuum filter small volumes for particulate or biological contamination analysis.

$355.10 - $375.00

Specifications

Autoclavable Not Autoclavable
Material Borosilicate Glass
Sterility Non-sterile
Filtration Area 2.5 cm2
Type Laboratory Filter Holders or Cyclones
View More Specs

Products 2
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
XX1012500
View Documents Promotion Details
MilliporeSigma™
XX1012500
With Glass frit membrane support
Each for $375.00
 
XX1012530
View Documents Promotion Details
MilliporeSigma™
XX1012530
With Stainless steel screen membrane support
Each for $355.10
 
Description

Description

Vacuum filter small volumes for particulate or biological contamination analysis.

 

  • Funnel capacity: 15mL
  • Filtration area: 2.5cm2
  • Not autoclavable with filter in place

 

Specifications

Specifications

Not Autoclavable
Non-sterile
Laboratory Filter Holders or Cyclones
6 in.
0.98 in.
Borosilicate Glass
2.5 cm2
15 mL
15.2 cm
2.5 cm
SDS
Product Certifications